BROWN YELLOW BY1 1 * 100 mL
Catalog # REGEEPBY101
Supplier: REAGECON
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals