ContactUsLinkComponent
IOne 1250 Controller with Left Sided Configuration. Includes 4 Pumps and Touch Screen Display. Does Not Included Gas Control, Vessel, or Probes. 115V 1 * 1 items
Catalog # DISI2022-8101
Supplier:  DISTEK
IOne 1250 Controller with Left Sided Configuration. Includes 4 Pumps and Touch Screen Display. Does Not Included Gas Control, Vessel, or Probes. 115V 1 * 1 items
Catalog # DISI2022-8101
Supplier:  DISTEK
Frequently Bought Together