Orders LinkContactUsLinkComponent
Pointed Scissors
150 mm
Catalog # SAMPA364-150
Supplier:   Sampling Systems
Pointed Scissors
Catalog # SAMPA364-150
Supplier:  Sampling Systems
Pointed Scissors
Pointed Scissors
150 mm
Price
10,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Surgical grade scissors for cutting.
  • Made from surgical grade 420 stainless steel
  • Great for cutting and manipulating samples
These high quality pointed scissors are made from surgical grade 420 stainless steel and great for cutting and manipulating.
Specifications
  • Length:
    150 mm
  • Seq:
    01