Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NEBNext Multiplex Oligos for Illumina Set 3 (96 Unique Dual Index Primer Pairs) 1 * 96 Tests
Catalog # NEWEE6444S
Supplier:
NEW ENGLAND BIOLABS
Expand image
NEBNext Multiplex Oligos for Illumina Set 3 (96 Unique Dual Index Primer Pairs) 1 * 96 Tests
Catalog # NEWEE6444S
Supplier:
NEW ENGLAND BIOLABS
Frequently Bought Together
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection