0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Back box cable to display unit 1 * 1 items
AIRLACCELEC3
:
Air Liquide
Back box cable to display unit 1 * 1 items
AIRLACCELEC3
:
Air Liquide
Frequently Bought Together
G10 PRINTER COVER - LT GRAY 1 * 1 items
: Thermo Scientific
CAP BLUE MLINE 1 * 1 items
: Sartorius
REPL COVER for CELL LOCKER COMPL. 1 * 1 items
: Thermo Scientific
window 1 * 1 items
: CleanAir by Baker
Seal for working chamber, 2,20m, for model 500 1 * 1 items
: MEMMERT
BIOTECTOR BASE REAGENT 1 * 20 L
: Hach
HSG ASSY PUMP UPPER W/BEARING 1 * 1 items
: Thermo Scientific
FAN MOTOR 1 * 1 items
: Binder
Accessories for Research® Plus pipettes
: EPPENDORF
Counter for volume display 2,5µl, 20µl, 200µl, 300µl 1 * 1 items
: EPPENDORF