ContactUsLinkComponent
Eluent concentrate for IC: 0.45M Na2CO3 0.14M NaHCO3 in H2O Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
Catalog # CMPLEL009.W.L5
Eluent concentrate for IC: 0.45M Na2CO3 0.14M NaHCO3 in H2O Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
Catalog # CMPLEL009.W.L5
Frequently Bought Together