UPLFLN100XO2-2 Objective Universal Plan Fluorite oil immersion objective with 100x magnification. With working distance of 0.2 mm, numerical aperture of 1.3 and cover correction of 0.17 mm. 1 * 1 ST
Catalog # OLYMN5703400
Supplier: OLYMPUS OPTICAL
Frequently Bought Together