ContactUsLinkComponent
BX53F2 LED Set containing BX53F2 and U-LHLEDC100. 1. BX53F2 Upright manual microscope frame for reflected and transmitted light observations. Prefocusing limit stopper and torque adjustment on coarse focus. Front side mounted light intensity control 1 * 1 ST
Catalog # OLYME0432197S
Supplier:  OLYMPUS OPTICAL
BX53F2 LED Set containing BX53F2 and U-LHLEDC100. 1. BX53F2 Upright manual microscope frame for reflected and transmitted light observations. Prefocusing limit stopper and torque adjustment on coarse focus. Front side mounted light intensity control 1 * 1 ST
Catalog # OLYME0432197S
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together