Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP 1 * 25 kg
Catalog # SPCMSO130-25KGBL
Supplier: Spectrum Chemical
Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP 1 * 25 kg
Catalog # SPCMSO130-25KGBL
Supplier: Spectrum Chemical
Frequently Bought Together
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium sulphite 95.0-100.5%, purified
Supplier: VWR Chemicals
Iodine ≥99%, GPR RECTAPUR®, resublimed
Supplier: VWR Chemicals
Potassium chlorate, EMSURE® for analysis, Supelco®
Supplier: Merck
Silver nitrate ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
Supplier: VWR Chemicals
Potassium fluoride ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Calcium chloride hexahydrate ≥97%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium carbonate, anhydrous ≥99% ACS
Supplier: VWR Chemicals
Potassium hydrogen carbonate, EMSURE® for analysis, Supelco®
Supplier: Merck
Dipotassium-L(+)-tartrate hemihydrate ≥99%
Supplier: Thermo Fisher Scientific
Barium carbonate 99.0-101.0% ACS
Supplier: Thermo Fisher Scientific
Aluminium chloride, anhydrous 98.5%, powder, extra pure
Supplier: Thermo Fisher Scientific
Sodium chlorate 99% ACS
Supplier: Thermo Fisher Scientific