STANDARD AU 2MG/L IN HNO3 5% HCL 15% 1 * 500 mL
Catalog # CMPLRM087308L5.2
Supplier: CUSTOM MADE CHEMICALS LAB
STANDARD AU 2MG/L IN HNO3 5% HCL 15% 1 * 500 mL
Catalog # CMPLRM087308L5.2
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
TRIGLYCINE 1 * 100 mg
Supplier: Merck
Acetaldehyde ≥95%
Supplier: LGC Standards PROMOCHEM
Propylbenzene, Supelco®
Supplier: Merck
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
Supplier: YSI ENVIRONMENTAL
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Dimethoate certified 1 * 0,1 g
Supplier: EHRENSTORFER
alpha-HCH D6 certified 1 * 10 mg
Supplier: EHRENSTORFER
METHYL 3-AMINO-4-METHYLTHIOPHENE-2-CARBO 1 * 100 mg
Supplier: Merck
1,1-Dichloro-2,2-bis(4-ethylphenyl)ethane
Supplier: EHRENSTORFER
Chlorobenzene-[D5] 2,000 µg/ml in methanol, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Luteolin 7-glucoside
Supplier: Merck
4-ISOPROPYLPHENOL 1 * 50 mg
Supplier: Merck