Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD AU 0.5 MG/L IN HNO3 5% HCL 15% 1 * 500 mL
Catalog # CMPLRM087306L5.2
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
STANDARD AU 0.5 MG/L IN HNO3 5% HCL 15% 1 * 500 mL
Catalog # CMPLRM087306L5.2
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, ready to use, 1 litre bottles, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach