REAGECON OSMOLALITY STANDARD 500 MOSM/KG 1 * 100 mL
Catalog # REGERE-OSM50001
Supplier: REAGECON
Frequently Bought Together
Sucrose, Reference Standard
Supplier: USP
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
Conductivity standards
Supplier: HAMILTON BONADUZ
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Ethanolamine, Supelco®
Supplier: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
N-Nitroso-ethyl methylamine 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
METHACRYLIC ACID-ETHYLACRYLATE COPOLYMER 1 * 1 items
Supplier: EDQM