REAGECON OSMOLALITY STANDARD 100 MOSM/KG 1 * 100 mL
Catalog # REGERE-OSM10001
Supplier: REAGECON
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Mepronil certified 1 * 0,1 g
Supplier: EHRENSTORFER
BTEX Mixture 884 200 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
PRUDOMESTIN, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
Tris(2-chloro-1-(chloromethyl)ethyl) phosphate certified 1 * 0,25 g
Supplier: EHRENSTORFER
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
Polystyrene, analytical standards for GPC
Supplier: Merck
CEFAZOLIN SODIUM SALT 1 * 75 mg
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
Elemental analysis standards, sulphur standards, VHG
Supplier: LGC Standards PROMOCHEM
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
AMOXICILLIN TRIHYDRATE PERFORMANCE VERIF
Supplier: EDQM
Dysprosium standard solution, 1000 mg/L Dy in 5% nitric acid (from Dy₂O₃), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Milrinone, Reference Standards
Supplier: USP
pH buffer solutions, Specpure®
Supplier: Thermo Fisher Scientific