Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
20ML PL TOTAL HARDNESS 1 (50 TESTS) 1 * 20 mL
Catalog # WTIDPOL20TH1
Supplier:
WATER ID PRIMELAB
Expand image
20ML PL TOTAL HARDNESS 1 (50 TESTS) 1 * 20 mL
Catalog # WTIDPOL20TH1
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
Fast blue BB salt ≥80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Araldite® M, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
Eosin Y (yellowish) 5 wt. % in H₂O in aqueous solution, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Decalcifier, rapid, histograde, for diagnostics use 1 * 2,5 L
Supplier: Avantor
Stain,Hematoxylin Gill 1,4x1PT 1 * 4 items
Supplier: Epredia