REAGENT KIT FOR PRIMELAB ID68(541 TESTS) 1 * 1 items
Catalog # WTIDPL68-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID68(541 TESTS) 1 * 1 items
Catalog # WTIDPL68-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test strips, quaternary ammonium compounds (QUAT), QUANTOFIX®
Supplier: MACHEREY-NAGEL
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Joint grease, K.W.S.
Supplier: POLLATH
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
Sodium hypochlorite 0.5 - 0.7% available chlorine, J.T.Baker®
Supplier: Avantor
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
1-Hexanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach