REAGENT KIT FOR PRIMELAB ID34(162 TESTS) 1 * 1 items
Artikelnr. WTIDPL34-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID34(162 TESTS) 1 * 1 items
Artikelnr. WTIDPL34-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for semi-quantitative analysis of low water hardness, AQUADUR® sensitive
Supplier: MACHEREY-NAGEL
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, nitrite, Dosatest
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach