REAGENT KIT FOR PRIMELAB ID150(50 TESTS) 1 * 1 items
WTIDPL150-KIT
Frequently Bought Together
Ammonia solution ∼32%, GPR RECTAPUR®
: VWR Chemicals
Sodium hypochlorite (14% Cl₂) in aqueous solution, TECHNICAL
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Test strips, peroxide, Dosatest
: VWR Chemicals
Powder pillows, ammonia
: Hach