ContactUsLinkComponent
Phenol CAS:108-95-2 EC:203-632-7 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 250 g
Catalog # CMPLSB4103.250G
Phenol CAS:108-95-2 EC:203-632-7 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 250 g
Catalog # CMPLSB4103.250G
Frequently Bought Together