ContactUsLinkComponent
Masterflex® Ismatec® Pressure lever cassette with inserts PVDF, 4/pk
Masterflex® Ismatec® Pressure lever cassette with inserts PVDF, 4/pk
Catalog # MFLX78001-97
Masterflex® Ismatec® Pressure lever cassette with inserts PVDF, 4/pk
Catalog # MFLX78001-97
About this item
Choose new cassettes based upon material of construction, tubing type and pressure capabilities.
  • Material of construction standard POM-C or optional PVDF
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.
  • Description:
    Pressure lever cassette with inserts PVDF, 4/pk
  • Seq:
    10
  • Environmentally Preferable:
  • Pk:
    4
Pressure lever cassettes with inserts
Product Information