1 component: Perfluorooctanoic acid [CAS:335-67-1] 100mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a freezer at -18C or below 1 * 1 mL
Catalog # CMPLRD0030681
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together