Orders LinkContactUsLinkComponent
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Labels, rectangular, B-461, M7-119-461, Polyester, Permanent acrylic, White/Transparent, M710, 15,24W×48,26H mm, 500/box
Catalog # BRDY311979
Supplier:   Brady
Self-laminating Polyester Labels for M710
Catalog # BRDY311979
Supplier:  Brady
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Labels, rectangular, B-461, M7-119-461, Polyester, Permanent acrylic, White/Transparent, M710, 15,24W×48,26H mm, 500/box
Price
253,00 €
Log in for availability
1 * 1 Rolle
Product Details & Documents
Self-laminating polyester labels for M710 printer.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Specifications
  • Printer model:
    M710
  • Type:
    B-461
  • Colour:
    White/Transparent
  • Description:
    Labels, rectangular
  • Dimensions:
    15,24W×48,26H mm
  • Material:
    Polyester
  • Seq:
    04
  • Model:
    M7-119-461
  • Packed:
    500/box
  • Adhesive:
    Permanent acrylic