Orders LinkContactUsLinkComponent
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Labels, rectangular and round, B-101, M7-101-461, Polyester, Permanent acrylic, White/Transparent, M710, 40,00W×15,00H×9,53Ø m, 250 sets/box
Catalog # BRDY311974
Supplier:   Brady
Self-laminating Polyester Labels for M710
Catalog # BRDY311974
Supplier:  Brady
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Labels, rectangular and round, B-101, M7-101-461, Polyester, Permanent acrylic, White/Transparent, M710, 40,00W×15,00H×9,53Ø m, 250 sets/box
Price
104,00 €
Log in for availability
1 * 250 SET
Product Details & Documents
Self-laminating polyester labels for M710 printer.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Specifications
  • Printer model:
    M710
  • Type:
    B-101
  • Colour:
    White/Transparent
  • Description:
    Labels, rectangular and round
  • Dimensions:
    40,00W×15,00H×9,53Ø m
  • Material:
    Polyester
  • Seq:
    09
  • Model:
    M7-101-461
  • Packed:
    250 sets/box
  • Adhesive:
    Permanent acrylic