Orders LinkContactUsLinkComponent
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Labels, rectangular and round, B-116, M7-116-461, Polyester, Permanent acrylic, White/Transparent, M710, 15,24W×41,28H×12,70Ø mm, 150 sets/box
Catalog # BRDY311977
Supplier:   Brady
Self-laminating Polyester Labels for M710
Catalog # BRDY311977
Supplier:  Brady
Self-laminating Polyester Labels for M710
Self-laminating Polyester Labels for M710
Labels, rectangular and round, B-116, M7-116-461, Polyester, Permanent acrylic, White/Transparent, M710, 15,24W×41,28H×12,70Ø mm, 150 sets/box
Price
64,90 €
Log in for availability
1 * 150 SET
Product Details & Documents
Self-laminating polyester labels for M710 printer.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.
Specifications
  • Printer model:
    M710
  • Type:
    B-116
  • Colour:
    White/Transparent
  • Description:
    Labels, rectangular and round
  • Dimensions:
    15,24W×41,28H×12,70Ø mm
  • Material:
    Polyester
  • Seq:
    08
  • Model:
    M7-116-461
  • Packed:
    150 sets/box
  • Adhesive:
    Permanent acrylic