ContactUsLinkComponent
1ml SQW Mikro-Insert, 45.9 x 7.6mm, clear glass, with assembled 9mm PE Cap, 5.9mm high, red, Silicone white/PTFE red, 45 shore A, 1.9mm, slitted 1 * 960 items
Catalog # ALUG1013340
Supplier:  APG Europe
1ml SQW Mikro-Insert, 45.9 x 7.6mm, clear glass, with assembled 9mm PE Cap, 5.9mm high, red, Silicone white/PTFE red, 45 shore A, 1.9mm, slitted 1 * 960 items
Catalog # ALUG1013340
Supplier:  APG Europe
Frequently Bought Together