TRICLABENDAZOLE FOR SYSTEM SUITABILITY 1 * 0,12 mg
Catalog # EDQMY0001723
Supplier: EDQM
Frequently Bought Together
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH buffer solutions, colour coded
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond