UNIVERSAL POWER ADAPTOR FOR CRYOLYS 1 * 1 items
Catalog # BERTS000845CLYS0A0
Supplier: Bertin Instruments
UNIVERSAL POWER ADAPTOR FOR CRYOLYS 1 * 1 items
Catalog # BERTS000845CLYS0A0
Supplier: Bertin Instruments
Frequently Bought Together
Cable set 230V 1 * 1 items
Supplier: MEMMERT
Eutectic point meter. 1 * 1 items
Supplier: Azbil Telstar Technologies
MEM SWTCH INCUBATG MINI SHAKER 1 * 1 items
Supplier: VWR Collection
EPROM 1 * 1 items
Supplier: Thermo Scientific
O RING PERFLUORO/FFKM SLIP RING RV16 FDA COMPLIANT 1 * 1 items
Supplier: HWS LABORTECHNIK
SPARES-KIT -T N96 1 * 1 items
Supplier: KNF
DOOR SEAL 692 X 641 MM, WITH 4-SIDED MAGNET, WHITE, SUCCESSOR OF: 687 X 632 MM, 1 * 1 items
Supplier: KIRSCH PHILIPP
Defrost valve solenoid 1 * 1 items
Supplier: LABCONCO
BLUE FILTER HOLDER FOR BA310 MET 1 * 1 items
Supplier: MOTIC
Accessories for multi channel pipettes, mechanical, Research® plus
Supplier: EPPENDORF
NEW ORING 100 SET- SINGLE (ALL) 1 * 10 items
Supplier: VWR Collection
front panel RO 5 1 * 1 items
Supplier: IKA
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
spare shield R-220 1 * 1 items
Supplier: BUCHI