Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

40...300 °C, 255 °C, 245 °C, Clear, 5 L
Catalog # JULA8940113
Supplier:   JULABO GmbH
Bath fluids
Catalog # JULA8940113
Supplier:  JULABO GmbH
CAS Number:  
Bath fluids
Bath fluids

40...300 °C, 255 °C, 245 °C, Clear, 5 L

CAS Number:  
Price
390,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.

  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness
Specifications

Specifications

  • Pk:
    5 L
  • Colour:
    Clear
  • Fire point:
    255 °C
  • Flash point:
    245 °C
  • Temperature range:
    40...300 °C

Specifications