HG 0.05MG/L IN HNO3 2% 1 * 100 mL
Catalog # CMPLRM038670L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, Nitrate, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Sodium hydroxide 30% in water, Technical Grade
Supplier: Thermo Fisher Scientific
Formalin 10%, buffered (neutral) (pH 7.0 ± 0.2), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Accessories for Technovit® 9100
Supplier: HERAEUS KULZER