Order Entry
Austria
Orders LinkContactUsLinkComponent
(S)-(-)-2-Methoxypropionic acid 97%

(S)-(-)-2-Methoxypropionic acid 97%

Bottle, 25 g
Catalog # APOSOR923407-25G
Supplier:   Apollo Scientific
(S)-(-)-2-Methoxypropionic acid 97%
Catalog # APOSOR923407-25G
Supplier:  Apollo Scientific
CAS Number:  23953-00-6
(S)-(-)-2-Methoxypropionic acid 97%

Bottle, 25 g

CAS Number:  23953-00-6
Price
417,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (S)-2-Methoxypropanoic acid
  • CAS Number:23953-00-6
  • Formula:C₄H₈O₃
  • MDL Number:MFCD01632589
  • Molecular Weight:104.105 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications