ContactUsLinkComponent
Masterflex® Sanitary Gasket, Buna-N, Flanged, 2" Tri-Clamp®; 10/PK
Masterflex® Sanitary Gasket, Buna-N, Flanged, 2" Tri-Clamp®; 10/PK
Catalog # MFLX30564-46
Masterflex® Sanitary Gasket, Buna-N, Flanged, 2" Tri-Clamp®; 10/PK
Catalog # MFLX30564-46
About this item
Hands-free installation for difficult-to-reach locations!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Material: Buna-N
  • Clamp Size: 2"
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port - once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time.
Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).
  • Sanitary clamp size (Port 1):
    2"
  • Connection type:
    Sanitary
  • Description:
    Sanitary Gasket, Buna-N, Flanged, 2" Tri-Clamp®; 10/PK
  • Environmentally Preferable:
  • Pk:
    10
  • Fitting material:
    Buna-N
  • Sanitary gasket size:
    2"
  • Warranty:
    1 year
Frequently Bought Together