SODIUM HYDROXIDE SOLUTION 0.1N BIOCERT 1 * 25 L
Catalog # SPCMS9291-25LT
Supplier: Spectrum Chemical
SODIUM HYDROXIDE SOLUTION 0.1N BIOCERT 1 * 25 L
Catalog # SPCMS9291-25LT
Supplier: Spectrum Chemical
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Joint grease, K.W.S.
Supplier: POLLATH
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
NANOCOLOR® Photometric Tube Test Kits for Phosphates (Ortho) and Phosphates (Total)
Supplier: MACHEREY-NAGEL
Visual colour kits, chlorine and pH, MQuant®, Supelco®
Supplier: Merck
Test strips for nickel, MQuant®, Supelco®
Supplier: Merck
Chlorine, sensitive test strips, Dosatest
Supplier: VWR Chemicals
Test kits, chlorine, Spectroquant®, Supelco®
Supplier: Merck
Test strips for glucose, MQuant®, Supelco®
Supplier: Merck
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, photometric, Chromium, LCK cuvette tests
Supplier: Hach
RP500 MCART LAC 250 TEST 1 * 250 Tests
Supplier: BAYER VITAL
Crystal violet solution
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond