BROMOMETHAN 2000 MG/L IN METHANOL 5 X1ML 1 * 5 mL
Catalog # CMPLRD0058571.5
Supplier: CUSTOM MADE CHEMICALS LAB
BROMOMETHAN 2000 MG/L IN METHANOL 5 X1ML 1 * 5 mL
Catalog # CMPLRD0058571.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach