ContactUsLinkComponent
The Leica DM6 B is an automated upright microscope with motorized Z-focus drive, fully automated transmitted light axis with perfectly integrated LED illumination for a constant color temperature. The integrated Illumination Manager automatically set 1 * 1 items
Catalog # LEIM11889186
Supplier:  LEICA MICROSYSTEMS
The Leica DM6 B is an automated upright microscope with motorized Z-focus drive, fully automated transmitted light axis with perfectly integrated LED illumination for a constant color temperature. The integrated Illumination Manager automatically set 1 * 1 items
Catalog # LEIM11889186
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together