class 1000, 100 x 130 x 0,10 mm 1 * 25 items
Catalog # BSNC101308981
Supplier: Basan
Frequently Bought Together
Cover spectacles, uvex super OTG
Supplier: UVEX
Autoclave tapes
Supplier: PRECISION DYNAMICS TIMEM
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
VWR®, Cleanroom Face Masks, Class 100
Supplier: VWR Collection
VWR® Amsterdam, Safety Spectacles
Supplier: VWR Collection
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
VWR®, Ladies' Coats, Cotton or Polyester/Cotton
Supplier: VWR Collection
Labelling tapes, temperature resistant
Supplier: PRECISION DYNAMICS TIMEM
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Supplier: Ansell
Colour Lab Tapes
Supplier: GA
VWR®, Men's Coats, Cotton or Polyester/Cotton
Supplier: VWR Collection
Labels, hazardous substance, GHS pictograms
Supplier: Brady
VWR®, Permanent Marker Pens
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-500
Supplier: Ansell