(E)-4-Bromobut-2-enoic acid,95% 1 * 10 g
Catalog # AMBDA698521-10G
Supplier: AMBEED
Frequently Bought Together
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Sinapic acid, Supelco®
Supplier: Merck
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane
Supplier: Roth Carl
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck