(3-Fluoro-4-(methoxy(methyl)carbamoyl)phenyl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA518965-250MG
Supplier: AMBEED
(3-Fluoro-4-(methoxy(methyl)carbamoyl)phenyl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA518965-250MG
Supplier: AMBEED
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor