3-Bromo-2-methoxybenzaldehyde,95% 1 * 5 g
Catalog # AMBDA105276-5G
Supplier: AMBEED
Frequently Bought Together
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hexane anhydrous, 95 % 1 * 2 L
Supplier: Merck
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium fluoride 99.9%, extra pure
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Glycerine (synthetic) 99+%, pure
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
1,2-DIMETHOXYETHANE 1 * 1 L
Supplier: Merck
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Diisopropyl ether, Sigma-Aldrich®
Supplier: Merck
n-Hexane
Supplier: Roth Carl
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl