N-Benzyl-N,N-dimethyl-3-tetradecanamidopropan-1-aminium chloride,98% 1 * 5 g
Catalog # AMBDA168180-5G
Supplier: AMBEED
N-Benzyl-N,N-dimethyl-3-tetradecanamidopropan-1-aminium chloride,98% 1 * 5 g
Catalog # AMBDA168180-5G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
PETROLEUM ETHER 40-60°C 1 * 5 L
Supplier: Roth Carl
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem