Potassium trifluoro(3-methylthiophen-2-yl)borate,95% 1 * 0,25 g
Catalog # AMBDA207216-250MG
Supplier: AMBEED
Potassium trifluoro(3-methylthiophen-2-yl)borate,95% 1 * 0,25 g
Catalog # AMBDA207216-250MG
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
1-Butanol NF
Supplier: Spectrum Chemical
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Methanol ROTISOLV® = 99,98%, Ultra LC-MS, 2.5L 1 * 2,5 L
Supplier: Roth Carl
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
TOLUOL CHROMASOLV FUER DI 1 * 18 L
Supplier: Honeywell Chemicals
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1,5-Diaminopentane 98%
Supplier: Thermo Fisher Scientific