4-(Trifluoromethoxy)toluene,98% 1 * 5 g
Catalog # AMBDA760913-5G
Supplier: AMBEED
Frequently Bought Together
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
2-Propanol, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor