1,3-Bis(3,5-bis(trifluoromethyl)phenyl)urea,98% 1 * 0,1 g
Catalog # AMBDA383673-100MG
Supplier: AMBEED
1,3-Bis(3,5-bis(trifluoromethyl)phenyl)urea,98% 1 * 0,1 g
Catalog # AMBDA383673-100MG
Supplier: AMBEED
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
1,2-Dichloroethane, Sigma-Aldrich®
Supplier: Merck
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals