3,4-Dichlorophenylhydrazine hydrochloride,98% 1 * 5 g
Catalog # AMBDA195716-5G
Supplier: AMBEED
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Bromine 99.6% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol technical grade 1 * 1.000 mL
Supplier: PanReac AppliChem
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isobutanol 99+% ACS
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
1,4-Dioxane, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99% (by GC) stabilised, Puriss., meets analytical specification of Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane 99+%, pure
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific