Sodium diethyl oxalacetate,95% 1 * 25 g
Catalog # AMBDA247452-25G
Supplier: AMBEED
Frequently Bought Together
Methylcyclohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Water for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck