Methyl 2-amino-4,5-difluorobenzoate,98% 1 * 25 g
AMBDA147427-25G
: AMBEED
Frequently Bought Together
1-Propanol, EMPLURA®, Supelco®
: Merck
Isoamyl alcohol, TECHNICAL
: VWR Chemicals
n-Octane ≥98%
: Thermo Fisher Scientific
2-Methylbutane for HPLC
: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
: Thermo Fisher Scientific
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Water ACS, Deionized
: Thermo Fisher Scientific