H-Tle-OMe.HCl,97% 1 * 25 g
AMBDA190165-25G
: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
: Merck
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
: PanReac AppliChem
Ethanolamine ACS
: Thermo Fisher Scientific
8 components: 100mg/l each of Al Cr Cu Fe Mn Ba Zn Zr in HNO3 5% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
2,2,4-Trimethylpentane 99.5% for analysis
: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
n-Hexane
: Roth Carl
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
N-Methyl-2-pyrrolidone (NMP) 98%, pure
: Thermo Fisher Scientific