H-D-Ala-OH,97% 1 * 25 g
Catalog # AMBDA109670-25G
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Methyl acetate anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Isoamyl alcohol mixture of isomers ≥99%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform ≥99% stabilised ethanol-free
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals