4-Chloro-3-(trifluoromethyl)phenylisocyanate,97% GC 1 * 1 g
Catalog # AMBDA618012-1G
Supplier: AMBEED
4-Chloro-3-(trifluoromethyl)phenylisocyanate,97% GC 1 * 1 g
Catalog # AMBDA618012-1G
Supplier: AMBEED
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Hexane anhydrous, 95 % 1 * 250 mL
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck