Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate,95+% 1 * 0,1 g
AMBDA750602-100MG
: AMBEED
Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate,95+% 1 * 0,1 g
AMBDA750602-100MG
: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
: Honeywell Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Dimethyl sulphoxide, anhydrous ≥99.8%
: Thermo Fisher Scientific
Nitrobenzene 99+% ACS
: Thermo Fisher Scientific