1-Benzyl-3-iodo-1H-pyrrolo[2,3-b]pyridine,98+% 1 * 0,25 g
Catalog # AMBDA501449-250MG
Supplier: AMBEED
1-Benzyl-3-iodo-1H-pyrrolo[2,3-b]pyridine,98+% 1 * 0,25 g
Catalog # AMBDA501449-250MG
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Diisopropyl ether 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Sinapic acid, Supelco®
Supplier: Merck
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclopentane 95+%, pure
Supplier: Thermo Fisher Scientific
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
1-Butanol 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Water for LC-MS 1 * 2,5 L
Supplier: PanReac AppliChem