1-(Bromomethyl)naphthalene,97% 1 * 10 g
Catalog # AMBDA664482-10G
Supplier: AMBEED
Frequently Bought Together
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 0.5 M in HPLC grade water for HPLC
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
1-(Trimethylsilyl)imidazole, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific